Learn More
LICORbio Intercept (TBS) Blocking Buffer and Diluent Kit
Catalog No. NC1679011
Encompass_Preferred
Try Intercept (TBS) Blocking Buffer and Intercept T20 (TBS) Antibody Diluent for improved results in your quantitative Western blots and other immunoassays. Intercept Blocking Buffers provide excellent blocking performance with low background and low variability. Intercept T20 Antibody Diluents improve the specificity of the primary and secondary antibodies, reducing off-target effects. They are preformulated with Tween 20. There’s no need to mix the diluent yourself, which saves you time and reduces potential variation. This kit includes Intercept (TBS) Blocking Buffer (1 x 500 mL) and Intercept T20 (TBS) Antibody Diluent (2 x 500 mL)
Catalog No. NC1679011 Supplier LICORbio Supplier No. 92766003
Try Intercept (TBS) Blocking Buffer and Intercept T20 (TBS) Antibody Diluent for improved results in your quantitative Western blots and other immunoassays. Intercept Blocking Buffers provide excellent blocking performance with low background and low variability. Intercept T20 Antibody Diluents improve the specificity of the primary and secondary antibodies, reducing off-target effects. They are preformulated with Tween 20. There’s no need to mix the diluent yourself, which saves you time and reduces potential variation. This kit includes Intercept (TBS) Blocking Buffer (1 x 500 mL) and Intercept T20 (TBS) Antibody Diluent (2 x 500 mL)