Learn More
LICORbio Intercept (PBS) Blocking Buffer (3 x 500 mL)
Catalog No. NC1660562
Encompass_Preferred
Intercept (PBS) Blocking Buffers are formulated to provide highly efficient blocking and low background variability for replicable quantitative Western blots and other immunoassays. Intercept (PBS) Blocking Buffer is the blocking buffer of choice for near-infrared fluorescent applications when using a phosphate-buffered saline (PBS) system. It contains no mammalian proteins for the lowest cross-reactivity with mammalian antibodies, it is an excellent choice for biotin-streptavidin detection and it is suitable for a wide range of applications, including Western blots, In-Cell Western™ assays, and protein arrays. Intercept (PBS) Blocking Buffer is a ready-to-use blocker formulation in PBS. It contains no sodium azide, making it suitable for both near-infrared or chemiluminescent detection. It is stored at 4 °C and is also available in kits with Immobilon-FL PVDF and nitrocellulose membranes.
Catalog No. NC1660562 Supplier LICORbio Supplier No. 92770003
Intercept (PBS) Blocking Buffers are formulated to provide highly efficient blocking and low background variability for replicable quantitative Western blots and other immunoassays. Intercept (PBS) Blocking Buffer is the blocking buffer of choice for near-infrared fluorescent applications when using a phosphate-buffered saline (PBS) system. It contains no mammalian proteins for the lowest cross-reactivity with mammalian antibodies, it is an excellent choice for biotin-streptavidin detection and it is suitable for a wide range of applications, including Western blots, In-Cell Western™ assays, and protein arrays. Intercept (PBS) Blocking Buffer is a ready-to-use blocker formulation in PBS. It contains no sodium azide, making it suitable for both near-infrared or chemiluminescent detection. It is stored at 4 °C and is also available in kits with Immobilon-FL PVDF and nitrocellulose membranes.