Learn More
LICORbio Intercept T20 (PBS) Protein-Free Antibody Diluent (500mL)
Catalog No. NC2216067
Encompass_Preferred
Intercept T20 (PBS) Protein-Free Antibody Diluent improves the specificity of the primary and secondary antibodies, reducing off-target effects. The diluent contains Intercept (PBS) Protein-Free Blocking Buffer preformulated with Tween 20 for use in a phosphate-buffered saline (PBS) system. This diluent can be used where animal-based products are prohibited. Since there’s no need to mix the diluent yourself, it saves you time and reduces potential variation. Intercept T20 (PBS) Protein-Free Antibody Diluent contains a 0.05% concentration of Tween 20. It contains no sodium azide and is stored at 4 °C.
This item is not returnable. View return policy
Catalog No. NC2216067 Supplier LICORbio Supplier No. 92795001
Intercept T20 (PBS) Protein-Free Antibody Diluent improves the specificity of the primary and secondary antibodies, reducing off-target effects. The diluent contains Intercept (PBS) Protein-Free Blocking Buffer preformulated with Tween 20 for use in a phosphate-buffered saline (PBS) system. This diluent can be used where animal-based products are prohibited. Since there’s no need to mix the diluent yourself, it saves you time and reduces potential variation. Intercept T20 (PBS) Protein-Free Antibody Diluent contains a 0.05% concentration of Tween 20. It contains no sodium azide and is stored at 4 °C.