Learn More
LICORbio Intercept T20 (TBS) Antibody Diluent (3 x 500)
Catalog No. NC2216056
Encompass_Preferred
Intercept T20 (TBS) Antibody Diluent improves the specificity of the primary and secondary antibodies, reducing off-target effects. The diluent contains Intercept (TBS) Blocking Buffer in Tris-buffered saline (TBS) preformulated with Tween 20. Since there’s no need to mix the diluent yourself, it saves you time and reduces potential variation. Intercept T20 (TBS) Antibody Diluent contains a 0.05% concentration of Tween 20. It contains no sodium azide and is stored at 4 °C.
This item is not returnable. View return policy
Catalog No. NC2216056 Supplier LICORbio Supplier No. 92765003
Intercept T20 (TBS) Antibody Diluent improves the specificity of the primary and secondary antibodies, reducing off-target effects. The diluent contains Intercept (TBS) Blocking Buffer in Tris-buffered saline (TBS) preformulated with Tween 20. Since there’s no need to mix the diluent yourself, it saves you time and reduces potential variation. Intercept T20 (TBS) Antibody Diluent contains a 0.05% concentration of Tween 20. It contains no sodium azide and is stored at 4 °C.