Learn More
LICORbio Intercept T20 (TBS) Antibody Diluent (500 mL)
Catalog No. NC1703226
Encompass_Preferred
Intercept T20 (TBS) Antibody Diluent improves the specificity of the primary and secondary antibodies, reducing off-target effects. The diluent contains Intercept (TBS) Blocking Buffer in Tris-buffered saline (TBS) preformulated with Tween 20. Since there’s no need to mix the diluent yourself, it saves you time and reduces potential variation. Intercept T20 (TBS) Antibody Diluent contains a 0.05% concentration of Tween 20. It contains no sodium azide and is stored at 4 °C.
Catalog No. NC1703226 Supplier LICORbio Supplier No. 92765001
Intercept T20 (TBS) Antibody Diluent improves the specificity of the primary and secondary antibodies, reducing off-target effects. The diluent contains Intercept (TBS) Blocking Buffer in Tris-buffered saline (TBS) preformulated with Tween 20. Since there’s no need to mix the diluent yourself, it saves you time and reduces potential variation. Intercept T20 (TBS) Antibody Diluent contains a 0.05% concentration of Tween 20. It contains no sodium azide and is stored at 4 °C.