Learn More
LICORbio Intercept (TBS) Blocking Buffer (500 mL)
Catalog No. NC1660550
Encompass_Preferred
Intercept (TBS) Blocking Buffers are formulated to provide highly efficient blocking and low background variability for replicable quantitative Western blots and other immunoassays. Intercept (TBS) Blocking Buffer is a ready-to-use blocker formulation in Tris-buffered saline (TBS). It contains no mammalian proteins for the lowest cross-reactivity with mammalian antibodies; it can be used with the widest range of targets for Western blot detection; it is ideal for primary antibodies that are optimized for Tris-based buffer systems; it can be used for detection of phosphorylated targets; it has superior performance for alkaline phosphatase detection; it is an excellent choice for biotin-streptavidin detection.Shake well before each use.
Catalog No. NC1660550 Supplier LICORbio Supplier No. 92760001
Intercept (TBS) Blocking Buffers are formulated to provide highly efficient blocking and low background variability for replicable quantitative Western blots and other immunoassays. Intercept (TBS) Blocking Buffer is a ready-to-use blocker formulation in Tris-buffered saline (TBS). It contains no mammalian proteins for the lowest cross-reactivity with mammalian antibodies; it can be used with the widest range of targets for Western blot detection; it is ideal for primary antibodies that are optimized for Tris-based buffer systems; it can be used for detection of phosphorylated targets; it has superior performance for alkaline phosphatase detection; it is an excellent choice for biotin-streptavidin detection.Shake well before each use.