Learn More
LICOR Intercept (TBS) Protein-Free Blocking Buffer (10x500mL)
Supplier: LICOR 92780010
Intercept (TBS) Protein-Free Blocking Buffers are formulated to provide highly efficient blocking and low background variability for replicable quantitative Western blots and other immunoassays where animal-based products are prohibited. Intercept (TBS) Protein-Free Blocking Buffer is a ready-to-use blocker formulation in Tris-buffered saline (TBS). It contains no proteins, for assays where animal-based products are prohibited, it can be used with the widest range of targets for Western blot detection, it is ideal for primary antibodies that are optimized for Tris-based buffer systems, it can be used for detection of phosphorylated targets, it has superior performance for alkaline phosphatase detection, and it is an excellent choice for biotin-streptavidin detection. Intercept (TBS) Protein-Free Blocking Buffer is a ready-to-use blocker formulation in TBS and is stored at 4 °C. It contains no sodium azide, making it suitable for both near-infrared or chemiluminescent detection.