Learn More
LICORbio Intercept (TBS) Protein-Free Blocking Buffer (500 mL)
Catalog No. NC1680069
Encompass_Preferred
Item is non returnable
Intercept (TBS) Protein-Free Blocking Buffers are formulated to provide highly efficient blocking and low background variability for replicable quantitative Western blots and other immunoassays where animal-based products are prohibited. Intercept (TBS) Protein-Free Blocking Buffer is a ready-to-use blocker formulation in Tris-buffered saline (TBS). It contains no proteins, for assays where animal-based products are prohibited, it can be used with the widest range of targets for Western blot detection, it is ideal for primary antibodies that are optimized for Tris-based buffer systems, it can be used for detection of phosphorylated targets, it has superior performance for alkaline phosphatase detection, and it is an excellent choice for biotin-streptavidin detection.
Catalog No. NC1680069 Supplier LICORbio Supplier No. 92780001
Item is non returnable
Intercept (TBS) Protein-Free Blocking Buffers are formulated to provide highly efficient blocking and low background variability for replicable quantitative Western blots and other immunoassays where animal-based products are prohibited. Intercept (TBS) Protein-Free Blocking Buffer is a ready-to-use blocker formulation in Tris-buffered saline (TBS). It contains no proteins, for assays where animal-based products are prohibited, it can be used with the widest range of targets for Western blot detection, it is ideal for primary antibodies that are optimized for Tris-based buffer systems, it can be used for detection of phosphorylated targets, it has superior performance for alkaline phosphatase detection, and it is an excellent choice for biotin-streptavidin detection.