Learn More
Pendotech Pendotech Pressuremat To Watson Marlow Interface Cable
Catalog No. NC1831915
Encompass
Interface cable for Watson Marlow pump - D25 connector to PressureMAT to start/stop function (6ft/2M)
Catalog No. NC1831915 Supplier Pendotech Supplier No. PMATCBWM25
Interface cable for Watson Marlow pump - D25 connector to PressureMAT to start/stop function (6ft/2M)