Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Interface cable with DB9 male/DB9 female connectors.
Catalog No. NC1652792
Encompass
Use these nine-pin interface cable assemblies with Masterflex L/S drives that have a nine-pin connector; including the 07550- and 07551-series computer-compatible drives. The cable is six feet (1.8 m) long.
Catalog No. NC1652792 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX2205054
Use these nine-pin interface cable assemblies with Masterflex L/S drives that have a nine-pin connector; including the 07550- and 07551-series computer-compatible drives. The cable is six feet (1.8 m) long.