Learn More
Mettler Toledo™ Interface Cables for DL31 Karl Fischer™ Titrators
Catalog No. 01911119 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Connector Types:
RS9 Female to Female
Catalog No. Connector Types
01-911-119 RS9 Female to Female
1 options
Catalog No. 01-911-119 Supplier Mettler Toledo™ Supplier No. 51190362
Add to Cart
Add to Cart
Accessory for use with DL31 Karl Fischer Titrators.
For use with DL31 Karl Fischer Titrators.
TRUSTED_SUSTAINABILITY
Specifications
Connector Types RS9 Female to Female
Type Cable
Length (English) 59.05 in.
Length (Metric) 1500 mm