Learn More
Inorganic Ventures 200.7 82 INTERFER CHK 4 125mL
SDP
Catalog No. 50723002
Encompass_Preferred
Inorganic Ventures' 200.7 Interference Check Standard 4 is a 125mL certified reference material set in a nitric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-002 Supplier Inorganic Ventures Supplier No. 2007ICS4125ML
Inorganic Ventures' 200.7 Interference Check Standard 4 is a 125mL certified reference material set in a nitric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search