Learn More
Inorganic Ventures 6020 INTERFER CHK SOLN A 500mL
SDP
Catalog No. 50723011
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Dilution: 1:10, Analyte: Al, K, Fe, S, Ca, Mg, P, and Na 1000 ug/mL each, C 2000 ug/mL, Mo and Ti 20 ug/mL each, and Cl- 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-011 Supplier Inorganic Ventures Supplier No. 6020ICS0A500ML
Matrix: Nitric Acid (HNO3), Dilution: 1:10, Analyte: Al, K, Fe, S, Ca, Mg, P, and Na 1000 ug/mL each, C 2000 ug/mL, Mo and Ti 20 ug/mL each, and Cl- 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search