Learn More
Cpi International Interferents Check Solution A | 500mL
SDP
Catalog No. NC0906907
Encompass
CPI International's Peak Performance Multi Element standards are produced in a controlled environment using ultra high-purity starting materials with NIST traceability and with ISO 17025 accordance. Each standard is verified instrumentally and gravimetrically and comes with a Certificate of Analysis and an SDS is available to download at sds.cpiinternational.com.
Catalog No. NC0906907 Supplier Cpi International Supplier No. 4400INTA1500
CPI International's Peak Performance Multi Element standards are produced in a controlled environment using ultra high-purity starting materials with NIST traceability and with ISO 17025 accordance. Each standard is verified instrumentally and gravimetrically and comes with a Certificate of Analysis and an SDS is available to download at sds.cpiinternational.com.