Learn More
Andwin Scientific INTERFERIN SIRNA TRANSFECT 1ML
SDP
Catalog No. NC9004235
Encompass_Preferred
The INTERFERin siRNA Transfection Reagent guarantee reproducible transfection, and attain over 90% silencing in a broad variety of cells. A 1 milliliter volume of this reagent is enough to perform 500 to 1000 transfections in 24-well plates. This product can be used with serum and antibiotics. It must be stored at 4C (39F). This product is sold individually in a quantity of 1 mL.
Catalog No. NC9004235 Supplier Andwin Scientific Supplier No. 89129930
The INTERFERin siRNA Transfection Reagent guarantee reproducible transfection, and attain over 90% silencing in a broad variety of cells. A 1 milliliter volume of this reagent is enough to perform 500 to 1000 transfections in 24-well plates. This product can be used with serum and antibiotics. It must be stored at 4C (39F). This product is sold individually in a quantity of 1 mL.