Learn More
Creative Biomart Recombinant Interferon Alfa 2a
Supplier:  Creative Biomart THP0078
Encompass_Preferred
Interferon a (human leukocyte protein moiety reduced). A type I interferon consisting of 165 amino acid residues with lysine in position 23. This protein is produced by recombinant DNA technology and resembles interferon secreted by leukocytes.
Catalog No. 50-208-4129