Learn More
Creative Biomart Interferon alpha 2b
Catalog No. 502084130
Encompass_Preferred
Interferon alpha 2b (human leukocyte clone hif-sn 206 protein moiety reduced). A type I interferon consisting of 165 amino acid residues with arginine in position 23. This protein is produced by recombinant DNA technology and resembles interferon secreted by leukocytes.
Catalog No. 50-208-4130 Supplier Creative Biomart Supplier No. THP0079
Add to Cart
Add to Cart
Interferon alpha 2b (human leukocyte clone hif-sn 206 protein moiety reduced). A type I interferon consisting of 165 amino acid residues with arginine in position 23. This protein is produced by recombinant DNA technology and resembles interferon secreted by leukocytes.