Learn More
Sigma Aldrich Fine Chemicals Biosciences Interferon alpha 2B human
Catalog No. 502904694
Encompass
The interferon 2 (IfN 2) exists in three forms 2a 2b and 2c. It consists of 165 amino acids and five helices. IfN 2B is found structurally stable at pH 7. The recombinant IfN is nonglycosylated compared to the natural human IfN .
Catalog No. 50-290-4694 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H616610UG
The interferon 2 (IfN 2) exists in three forms 2a 2b and 2c. It consists of 165 amino acids and five helices. IfN 2B is found structurally stable at pH 7. The recombinant IfN is nonglycosylated compared to the natural human IfN .