Learn More
Sino Biological Interferon alpha 2/IFNA2 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 13833MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Interferon alpha 2/IFNA2 (rh Interferon alpha 2/IFNA2; Catalog#13833-HNAY; P01563.1; Cys24-Glu188). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8716