Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon alpha-4, recombinant (mouse) | >95.0% | 50 UG
SDP

Catalog No. 5000290214
Encompass_Preferred

Recombinant mouse interferon alpha-4 produced in HEK293 cells with a C-terminal His tag. Supplied as a 50 μg vial of 0.22 μm filtered PBS (pH 7.4), purified and characterized by SDS-PAGE and COA; intended for research use in cell signaling and antiviral response assays. Store at -80°C and avoid repeated freeze-thaw cycles.

  • Expressed in HEK293 cells.
  • C-terminal His tag facilitates detection and purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Formulated in PBS, pH 7.4, and 0.22 μm filtered.
  • Endotoxin level <1 EU/μg.
  • Observed molecular weight ~22-27 kDa due to glycosylation.
  • Store at -80°C for long-term stability; avoid repeated freeze-thaw cycles.

Catalog No. 50-002-90214 Supplier Medchemexpress LLC Supplier No. HYP7640350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interferon alpha-4 produced in HEK293 cells with a C-terminal His tag. Supplied as a 50 μg vial of 0.22 μm filtered PBS (pH 7.4), purified and characterized by SDS-PAGE and COA; intended for research use in cell signaling and antiviral response assays. Store at -80°C and avoid repeated freeze-thaw cycles.

  • Expressed in HEK293 cells.
  • C-terminal His tag facilitates detection and purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Formulated in PBS, pH 7.4, and 0.22 μm filtered.
  • Endotoxin level <1 EU/μg.
  • Observed molecular weight ~22-27 kDa due to glycosylation.
  • Store at -80°C for long-term stability; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.