Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon alpha-4 recombinant (mouse) | 96.5% | 5 UG
SDP

Catalog No. 5000290033
Encompass_Preferred

Recombinant mouse interferon alpha-4 is a type I interferon with antiviral activity provided as a research-grade protein expressed in HEK293 mammalian cells and carrying a C-terminal His tag for purification and detection. The protein comprises amino acids C25-E186 (162 aa) and is supplied as a small, high-purity aliquot suitable for in vitro studies.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Contains 162 amino acids (C25-E186).
  • High purity (96.5%) suitable for research applications.
  • Supplied as a 5 μg research aliquot for in vitro use.

Catalog No. 50-002-90033 Supplier Medchemexpress LLC Supplier No. HYP764035UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interferon alpha-4 is a type I interferon with antiviral activity provided as a research-grade protein expressed in HEK293 mammalian cells and carrying a C-terminal His tag for purification and detection. The protein comprises amino acids C25-E186 (162 aa) and is supplied as a small, high-purity aliquot suitable for in vitro studies.

  • Recombinant protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Contains 162 amino acids (C25-E186).
  • High purity (96.5%) suitable for research applications.
  • Supplied as a 5 μg research aliquot for in vitro use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.