Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon alpha/beta receptor 1 Antibody (YA3833) | 110-130 kDa | 10 UL
SDP

Catalog No. 5000446701
Encompass_Preferred

This Interferon alpha/beta receptor 1 Antibody (YA3833) is a mouse-derived, non-conjugated IgG1 monoclonal antibody targeting Interferon alpha/beta receptor 1. It forms a heterodimeric receptor with IFNAR2 for type I interferons, activating the JAK-STAT signaling cascade to regulate interferon-responsive genes.

  • Mouse-derived monoclonal antibody
  • Non-conjugated IgG1 isotype
  • Targets interferon alpha/beta receptor 1
  • Reacts with human and mouse species
  • Suitable for WB, IHC-P, FC, and ELISA applications
  • Supplied as a liquid in PBS with 0.05% sodium azide
  • Purity is affinity purified
  • Store at -20°C for one year; avoid repeated freeze/thaw cycles

Catalog No. 50-004-46701 Supplier Medchemexpress LLC Supplier No. HYP8413610UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Interferon alpha/beta receptor 1 Antibody (YA3833) is a mouse-derived, non-conjugated IgG1 monoclonal antibody targeting Interferon alpha/beta receptor 1. It forms a heterodimeric receptor with IFNAR2 for type I interferons, activating the JAK-STAT signaling cascade to regulate interferon-responsive genes.

  • Mouse-derived monoclonal antibody
  • Non-conjugated IgG1 isotype
  • Targets interferon alpha/beta receptor 1
  • Reacts with human and mouse species
  • Suitable for WB, IHC-P, FC, and ELISA applications
  • Supplied as a liquid in PBS with 0.05% sodium azide
  • Purity is affinity purified
  • Store at -20°C for one year; avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.