Learn More
Medchemexpress LLC Interferon alpha/beta receptor 1 Antibody (YA3833) | Predicted band size: 64 kDa; Observed band size: 110-130 kDa | 100 UL
SDP
Catalog No. 5000449192
Encompass_Preferred
The Interferon alpha/beta receptor 1 Antibody (YA3833) is a mouse-derived, non-conjugated IgG1 monoclonal antibody designed to target Interferon alpha/beta receptor 1. It is supplied in PBS with 0.05% sodium azide and is affinity purified. This product is intended for research use only, supporting studies related to type I interferon signaling pathways.
  • Mouse host
  • Monoclonal antibody
  • Reactive with human and mouse species
  • Affinity purified
  • Non-conjugated
  • Isotype IgG1
  • Supplied as a liquid in PBS with 0.05% sodium azide
  • Stable for 1 year when stored at -20°C
  • Compatible with Western Blot, Immunohistochemistry-Paraffin, Flow Cytometry, and ELISA applications
Catalog No. 50-004-49192 Supplier Medchemexpress LLC Supplier No. HYP84136100UL
The Interferon alpha/beta receptor 1 Antibody (YA3833) is a mouse-derived, non-conjugated IgG1 monoclonal antibody designed to target Interferon alpha/beta receptor 1. It is supplied in PBS with 0.05% sodium azide and is affinity purified. This product is intended for research use only, supporting studies related to type I interferon signaling pathways.
  • Mouse host
  • Monoclonal antibody
  • Reactive with human and mouse species
  • Affinity purified
  • Non-conjugated
  • Isotype IgG1
  • Supplied as a liquid in PBS with 0.05% sodium azide
  • Stable for 1 year when stored at -20°C
  • Compatible with Western Blot, Immunohistochemistry-Paraffin, Flow Cytometry, and ELISA applications