Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon alpha/beta receptor 1 Antibody (YA3833) | Predicted band size: 64 kDa; Observed band size: 110-130 kDa | 50 UL
SDP

Catalog No. 5000446496
Encompass_Preferred

The Interferon alpha/beta receptor 1 Antibody (YA3833) is a mouse-derived, non-conjugated IgG1 monoclonal antibody that targets Interferon alpha/beta receptor 1. This antibody is intended for research use only. Interferon alpha/beta receptor 1 (IFNAR1), in conjunction with IFNAR2, forms the heterodimeric receptor crucial for type I interferon signaling. Binding of type I interferons initiates the JAK-STAT signaling cascade, which leads to the transcriptional regulation of interferon-regulated genes that mediate the interferon response.

  • Host: Mouse
  • Clonality: Monoclonal antibody
  • Species reactivity: Human, mouse
  • Conjugation: Non-conjugated
  • Isotype: IgG1
  • Appearance: Liquid
  • Formulation: Supplied in PBS with 0.05% sodium azide
  • Purity: Affinity purified
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Applications: WB, IHC-P, FC, ELISA

Catalog No. 50-004-46496 Supplier Medchemexpress LLC Supplier No. HYP8413650UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Interferon alpha/beta receptor 1 Antibody (YA3833) is a mouse-derived, non-conjugated IgG1 monoclonal antibody that targets Interferon alpha/beta receptor 1. This antibody is intended for research use only. Interferon alpha/beta receptor 1 (IFNAR1), in conjunction with IFNAR2, forms the heterodimeric receptor crucial for type I interferon signaling. Binding of type I interferons initiates the JAK-STAT signaling cascade, which leads to the transcriptional regulation of interferon-regulated genes that mediate the interferon response.

  • Host: Mouse
  • Clonality: Monoclonal antibody
  • Species reactivity: Human, mouse
  • Conjugation: Non-conjugated
  • Isotype: IgG1
  • Appearance: Liquid
  • Formulation: Supplied in PBS with 0.05% sodium azide
  • Purity: Affinity purified
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Applications: WB, IHC-P, FC, ELISA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.