Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Interferon alpha (Cell culture) ELISA Kit

Catalog No. 502741214
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-274-1214 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-274-1214 Supplier Invitrogen™ Supplier No. 411001
Add to Cart
Add to Cart

ELISA

The Human Interferon alpha A, 2, A/D, D, K and 4b (Hu IFNαA, IFNα2, IFNαA/D, IFNαD, IFNαK and IFNα4b ) ELISA quantitates Hu IFNα in human cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IFNα. Principle of the method The Human IFNα solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01562, P01563, P05013, P05014
Assay Range 12.5 to 500 pg/ml or 156 to 5000 pg/ml
Description Human IFN alpha ELISA Kit (Supernatant)
Conjugate HRP
Product Type ELISA
Sample Type Supernatant
Specificity No significant cross-reaction with human IFN-ß, IFN-g or IFN-w; mouse or rat IFN-a, IFN-ß or IFN-g; bovine IFN-t
Sufficient For 5 x 96 microplate wells
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3439, 3440
Gene Symbol IFL,IFN,IFN-ALPHA,IFN-alphaD3,IFNA@,IFNA1,IFN-alphaA,IFNAB,INFA2,IFNA2
Interassay CV <8%
Intraassay CV <8
Kit Contents Pre-coated 96 well Microplate, Detection Antibody Concentrate, Anti-IgG-HRP Concentrate, Concentrate Diluent, Standard, Wash Buffer Concentrate, Substrate Solution, Sample Diluent, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Measurement Absorbance at 450 nm
Precision Intra-assay CV ≤ 8%; inter-assay CV ≤ 8%
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Supernatant 100 μL
Storage Requirements 2°C to 8°C
Target IFN alpha
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 15 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.