Learn More
Cusabio Technology LLC interferon beta 1 fibroblast
Catalog No. 5000049149
Encompass_Preferred
The Human Interferon B (IFN-B/IFNB) ELISA Kit is designed to detect and quantify IFNB1 in various biological samples including serum urine cell culture supernatants tissue homogenates and cerebrospinal fluid (CSF) This sandwich-based assay offers a sensitivity of 7 8 pg/mL and a broad detection range from 31 25 pg/mL to 2000 pg/mL enabling precise quantitative analysis The procedure requires a sample volume of 50-100 L and can be completed within 1-5 hours with absorbance measurements taken at 450 nm This kit provides a reliable tool for researchers studying IFN-B s role in immune responses antiviral mechanisms or related clinical applications
Catalog No. 50-000-49149 Supplier Cusabio Technology LLC Supplier No. CSBE09889H
The Human Interferon B (IFN-B/IFNB) ELISA Kit is designed to detect and quantify IFNB1 in various biological samples including serum urine cell culture supernatants tissue homogenates and cerebrospinal fluid (CSF) This sandwich-based assay offers a sensitivity of 7 8 pg/mL and a broad detection range from 31 25 pg/mL to 2000 pg/mL enabling precise quantitative analysis The procedure requires a sample volume of 50-100 L and can be completed within 1-5 hours with absorbance measurements taken at 450 nm This kit provides a reliable tool for researchers studying IFN-B s role in immune responses antiviral mechanisms or related clinical applications