Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon beta Antibody (YA1560) | 10 UL
SDP

Catalog No. 5000440909
Encompass_Preferred

Interferon beta Antibody (YA1560) is a rabbit antibody reactive with human interferon beta, designed for laboratory use in applications like Western Blot (WB) and Immunoprecipitation (IP). It is supplied as a liquid formulation and is for research use only.

  • Host: Rabbit
  • Reactivity: Human
  • Appearance: Liquid
  • Concentration: 1 mg/mL
  • Conjugation: Non-conjugated
  • Applications: Western Blot (WB), Immunoprecipitation (IP)
  • Formulation: Supplied in 10 mM phosphate buffered saline, pH 7.4, 150 mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
  • Storage: Stored at -20°C for 1 year; avoid repeated freeze / thaw cycles.

Catalog No. 50-004-40909 Supplier Medchemexpress LLC Supplier No. HYP8181510UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Interferon beta Antibody (YA1560) is a rabbit antibody reactive with human interferon beta, designed for laboratory use in applications like Western Blot (WB) and Immunoprecipitation (IP). It is supplied as a liquid formulation and is for research use only.

  • Host: Rabbit
  • Reactivity: Human
  • Appearance: Liquid
  • Concentration: 1 mg/mL
  • Conjugation: Non-conjugated
  • Applications: Western Blot (WB), Immunoprecipitation (IP)
  • Formulation: Supplied in 10 mM phosphate buffered saline, pH 7.4, 150 mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
  • Storage: Stored at -20°C for 1 year; avoid repeated freeze / thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.