Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon beta Antibody (YA1560) | 100 UL
SDP

Catalog No. 5000441168
Encompass_Preferred

Interferon beta Antibody (YA1560) is a liquid laboratory chemical supplied by MedChemExpress for research use. It has a predicted and observed molecular weight of 22 kDa and is provided at a concentration of 1 mg/mL.

  • Host: Rabbit
  • Reactivity: Human
  • Clonality No.: YA1560
  • SwissProt ID: P01574
  • Gene ID: 3456
  • Concentration: 1 mg/mL
  • Conjugation: Non-conjugated
  • Application: WB, IP
  • Dilution Ratio: WB: 1:500-1:1000; IP: 1:50
  • Formulation: Supplied in Rabbit IgG in 10mM phosphate buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Shipping: Shipping with blue ice.
  • Identified uses: Laboratory chemicals, manufacture of substances.
  • Appearance: Liquid

Catalog No. 50-004-41168 Supplier Medchemexpress LLC Supplier No. HYP81815100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Interferon beta Antibody (YA1560) is a liquid laboratory chemical supplied by MedChemExpress for research use. It has a predicted and observed molecular weight of 22 kDa and is provided at a concentration of 1 mg/mL.

  • Host: Rabbit
  • Reactivity: Human
  • Clonality No.: YA1560
  • SwissProt ID: P01574
  • Gene ID: 3456
  • Concentration: 1 mg/mL
  • Conjugation: Non-conjugated
  • Application: WB, IP
  • Dilution Ratio: WB: 1:500-1:1000; IP: 1:50
  • Formulation: Supplied in Rabbit IgG in 10mM phosphate buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Shipping: Shipping with blue ice.
  • Identified uses: Laboratory chemicals, manufacture of substances.
  • Appearance: Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.