Learn More
Genscript Corporation IFN-γ, Mouse
Supplier:  Genscript Corporation Z02916100
Encompass
Sharing 41% sequence identity with human Interferon gamma (hIFN--γ), mouse IFN gamma (mIFN--γ)is a macrophage-activating factor.The active form of IFN--γ is an antiparallel dimer that sets off IFN--γ/JAK/STAT pathway. IFN--γ signaling does diverse biological functions primarily related to host defense and immune regulation, including antiviral and antibacterial defense, apoptosis, inflammation, and innate and acquired immunity.While IFN--γ–induced inflammatory cascade summons a variety of immune-related cell types, such as macrophages, natural killer (NK) cells and cytotoxic T lymphocytes (CTLs), IFN--γ is also implicated in resistance to NK cell and CTL responses and in immune escape in avariety of cancers.
Catalog No. 50-276-549