Learn More
Biotium Interferon gamma (IFNG)(IFNG/1598), CF405S conjugate, 0.1mg/mL
Catalog No. 5000658933
Encompass_Preferred
This antibody recognizes a protein of 20-25 kDa, identified as human interferon gamma (IFN-γ). Multiple forms exist due to variable glycosylation and under non-denaturing conditions due to dimers and tetramers. T lymphocytes and NK cells mainly produce IFN-γ. It is a pleiotropic cytokine involved in the regulation of nearly all phases of immune and inflammatory responses, including the activation, growth and differentiation of T cell, B cells, macrophages, NK cells and other cell types such as endothelial cells and fibroblasts. It has weak anti-viral and anti-proliferative activity, and potentiates the antiviral and anti-tumor effects of IFN-γ (type I interferon).Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blu
Catalog No. 50-006-58933 Supplier Biotium Supplier No. BNC041598100
Add to Cart
Add to Cart
This antibody recognizes a protein of 20-25 kDa, identified as human interferon gamma (IFN-γ). Multiple forms exist due to variable glycosylation and under non-denaturing conditions due to dimers and tetramers. T lymphocytes and NK cells mainly produce IFN-γ. It is a pleiotropic cytokine involved in the regulation of nearly all phases of immune and inflammatory responses, including the activation, growth and differentiation of T cell, B cells, macrophages, NK cells and other cell types such as endothelial cells and fibroblasts. It has weak anti-viral and anti-proliferative activity, and potentiates the antiviral and anti-tumor effects of IFN-γ (type I interferon).Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blu