Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon-gamma receptor 1 (human) Fc fusion protein | >95.0% | 50 UG
SDP

Catalog No. 5000293562
Encompass_Preferred

Recombinant human interferon-gamma receptor 1 (IFN-γ R1/CD119) fused to human Fc and expressed in HEK293 cells. Supplied lyophilized for use in biochemical and cell-based assays to study IFN-γ signaling and receptor interactions. High purity and recommended storage and reconstitution conditions support reliable research performance.

  • Expressed in HEK293 cells with a C-terminal human Fc fusion.
  • Contains amino acids E18-G245 (accession P15260-1).
  • Molecular weight approximately 60-75 kDa.
  • Purity greater than 95.0% by reducing SDS-PAGE.
  • Formulated lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Do not reconstitute to concentrations below 100 μg/mL in ddH2O.
  • Store lyophilized at -20°C; after reconstitution, stable at 4°C for 1 week; long-term storage at -20°C or -80°C recommended for aliquots.
  • Supplied as 50 μg lyophilized quantity.

Catalog No. 50-002-93562 Supplier Medchemexpress LLC Supplier No. HYP7485550UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interferon-gamma receptor 1 (IFN-γ R1/CD119) fused to human Fc and expressed in HEK293 cells. Supplied lyophilized for use in biochemical and cell-based assays to study IFN-γ signaling and receptor interactions. High purity and recommended storage and reconstitution conditions support reliable research performance.

  • Expressed in HEK293 cells with a C-terminal human Fc fusion.
  • Contains amino acids E18-G245 (accession P15260-1).
  • Molecular weight approximately 60-75 kDa.
  • Purity greater than 95.0% by reducing SDS-PAGE.
  • Formulated lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Do not reconstitute to concentrations below 100 μg/mL in ddH2O.
  • Store lyophilized at -20°C; after reconstitution, stable at 4°C for 1 week; long-term storage at -20°C or -80°C recommended for aliquots.
  • Supplied as 50 μg lyophilized quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.