Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon gamma receptor 1 (IFN-gamma R1) Fc-fusion protein | >95.0% | 5 UG
SDP

Catalog No. 5000292867
Encompass_Preferred

Recombinant human interferon-γ receptor 1 extracellular domain fused to a human Fc tag, expressed in HEK293 cells and supplied lyophilized for research use. The protein is intended for use in binding studies, biochemical characterization, and immunoassays requiring a soluble Fc-fusion receptor fragment.

  • Recombinant extracellular domain of IFN-γ receptor 1 fused to hFc.
  • Produced in HEK293 cells for human-like glycosylation.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Molecular weight ~60-75 kDa.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4 for stability.
  • Recommended storage at -20°C; reconstituted stability at 4°C for 1 week.
  • Suitable for binding assays and biochemical characterization.

Catalog No. 50-002-92867 Supplier Medchemexpress LLC Supplier No. HYP748555UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interferon-γ receptor 1 extracellular domain fused to a human Fc tag, expressed in HEK293 cells and supplied lyophilized for research use. The protein is intended for use in binding studies, biochemical characterization, and immunoassays requiring a soluble Fc-fusion receptor fragment.

  • Recombinant extracellular domain of IFN-γ receptor 1 fused to hFc.
  • Produced in HEK293 cells for human-like glycosylation.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Molecular weight ~60-75 kDa.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4 for stability.
  • Recommended storage at -20°C; reconstituted stability at 4°C for 1 week.
  • Suitable for binding assays and biochemical characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.