Learn More
Medchemexpress LLC Interferon-gamma receptor 1 (rat), Fc fusion protein | 100 UG
SDP
Catalog No. 5000292662
Encompass_Preferred
Recombinant rat interferon-gamma receptor 1 (CD119) extracellular domain fused to a human Fc tag, produced in HEK293 cells and supplied as a purified protein for in vitro research. The construct corresponds to residues M1-S241 and is suitable for receptor binding, signaling studies, and immunological assays. Supplied in a 100 μg format.
  • Rat origin extracellular domain (M1-S241) for species-relevant studies.
  • Expressed in HEK293 cells to support mammalian glycosylation.
  • C-terminal human Fc fusion for improved stability and detection.
  • Suitable for receptor binding and signaling assays.
  • Provided as a purified protein for in vitro research applications.
Catalog No. 50-002-92662 Supplier Medchemexpress LLC Supplier No. HYP75824100UG
Recombinant rat interferon-gamma receptor 1 (CD119) extracellular domain fused to a human Fc tag, produced in HEK293 cells and supplied as a purified protein for in vitro research. The construct corresponds to residues M1-S241 and is suitable for receptor binding, signaling studies, and immunological assays. Supplied in a 100 μg format.
  • Rat origin extracellular domain (M1-S241) for species-relevant studies.
  • Expressed in HEK293 cells to support mammalian glycosylation.
  • C-terminal human Fc fusion for improved stability and detection.
  • Suitable for receptor binding and signaling assays.
  • Provided as a purified protein for in vitro research applications.