Learn More
Medchemexpress LLC Interferon gamma receptor 1, rat, Fc fusion protein | >95.0% | 2 UG
SDP
Catalog No. 5000292215
Encompass_Preferred
Recombinant rat interferon-gamma receptor 1 (IFN-gamma R1/CD119) extracellular domain expressed in HEK293 cells and provided as an Fc fusion for enhanced stability. Intended for research use in immunology, receptor-ligand interaction studies, and assay development; supplied in microgram-scale sizes with recommended storage and handling for protein reagents.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • Fc tag for increased stability and dimerization.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in microgram-scale sizes suitable for assay development.
  • Suitable for receptor binding, biochemical assays, and antibody characterization.
  • Supplied with handling and storage guidance for protein reagents.
Catalog No. 50-002-92215 Supplier Medchemexpress LLC Supplier No. HYP758242UG
Recombinant rat interferon-gamma receptor 1 (IFN-gamma R1/CD119) extracellular domain expressed in HEK293 cells and provided as an Fc fusion for enhanced stability. Intended for research use in immunology, receptor-ligand interaction studies, and assay development; supplied in microgram-scale sizes with recommended storage and handling for protein reagents.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • Fc tag for increased stability and dimerization.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in microgram-scale sizes suitable for assay development.
  • Suitable for receptor binding, biochemical assays, and antibody characterization.
  • Supplied with handling and storage guidance for protein reagents.