Learn More
Medchemexpress LLC Interferon gamma receptor 1 (rat) Fc fusion protein | >95.0% | 50 UG
SDP
Catalog No. 5000290956
Encompass_Preferred
Recombinant rat interferon gamma receptor 1 extracellular domain Fc fusion protein expressed in HEK293 cells. Supplied lyophilized in a buffered formulation, it is intended for binding assays, receptor/ligand studies, and assay development that require the rat IFN-gamma receptor extracellular domain.
  • Expressed in HEK293 cells for mammalian glycosylation.
  • C-terminal human Fc fusion to facilitate detection and purification.
  • High purity (>95%) as verified by reducing SDS-PAGE.
  • Lyophilized formulation for extended shelf life.
  • Recommended reconstitution ≥100 μg/mL in ddH2O; store aliquots at -20°C or -80°C for long-term.
Catalog No. 50-002-90956 Supplier Medchemexpress LLC Supplier No. HYP7582450UG
Recombinant rat interferon gamma receptor 1 extracellular domain Fc fusion protein expressed in HEK293 cells. Supplied lyophilized in a buffered formulation, it is intended for binding assays, receptor/ligand studies, and assay development that require the rat IFN-gamma receptor extracellular domain.
  • Expressed in HEK293 cells for mammalian glycosylation.
  • C-terminal human Fc fusion to facilitate detection and purification.
  • High purity (>95%) as verified by reducing SDS-PAGE.
  • Lyophilized formulation for extended shelf life.
  • Recommended reconstitution ≥100 μg/mL in ddH2O; store aliquots at -20°C or -80°C for long-term.