Learn More
Sino Biological Interferon Gamma/IFN gamma/IFNG Antibody (HRP), Rabbit MAb 100ug
Supplier:  Sino Biological 11725R209H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Interferon Gamma/IFN gamma/IFNG (rh Interferon Gamma/IFN gamma/IFNG; Catalog#11725-HNAS; NP_000610.2; Met1-Gln166), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-7755