Learn More
Medchemexpress LLC HY-111669 10mg Medchemexpress, Sigma-2 receptor antagonist 1 CAS:1802632-22-9 Purity:>98%
SDP
Catalog No. 501968941
Encompass_Preferred
Medchemexpress, HY-112189 10mg Interferon receptor inducer-1 CAS:2215120-36-6 Interferon receptor inducer-1 (compound 6) is an interferon (IFN) receptor inducer. Used accordingly in the treatment of a disorder in which the induction of interferon is involved. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8941 Supplier Medchemexpress LLC Supplier No. HY11166910MG
Medchemexpress, HY-112189 10mg Interferon receptor inducer-1 CAS:2215120-36-6 Interferon receptor inducer-1 (compound 6) is an interferon (IFN) receptor inducer. Used accordingly in the treatment of a disorder in which the induction of interferon is involved. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.