Learn More
Invivogen HEK-Blue™ IFN-λ Cells, Detects Bioactive IFN-λs, Monitors JAK/ISGF3 Pathway, 1 vial
SDP
Catalog No. NC2543438
Encompass
The HEK-Blue™ IFN-λ Cells from InvivoGen are engineered human embryonic kidney (HEK293) cells designed for detecting bioactive human type III interferons (IFN-λs). These cells effectively monitor the JAK/ISGF3 pathway, providing a valuable tool for understanding IFN-λ signaling and screening potential inhibitors. This product stands out due to its specificity for human and murine IFN-λs, facilitating advanced research in therapeutic applications.
Catalog No. NC2543438 Supplier Invivogen Supplier No. HKBIFNLV2
The HEK-Blue™ IFN-λ Cells from InvivoGen are engineered human embryonic kidney (HEK293) cells designed for detecting bioactive human type III interferons (IFN-λs). These cells effectively monitor the JAK/ISGF3 pathway, providing a valuable tool for understanding IFN-λ signaling and screening potential inhibitors. This product stands out due to its specificity for human and murine IFN-λs, facilitating advanced research in therapeutic applications.