Learn More
MSC Interlake Mecalux Pallet Rack Beam, 48 in, 2.75 in, 6,050 lb
Catalog No. 5000530337
Encompass
The Interlake Mecalux Pallet Rack Beam provides a robust solution for industrial storage needs. It features a durable construction designed to support heavy loads in various pallet racking configurations.
  • 6,050 lb capacity
  • 0.172 in maximum deflection
  • 48 in width
  • 2.75 in overall height
  • Orange color
  • Made in the United States
Catalog No. 50-005-30337 Supplier MSC Supplier No. 66147216
The Interlake Mecalux Pallet Rack Beam provides a robust solution for industrial storage needs. It features a durable construction designed to support heavy loads in various pallet racking configurations.
  • 6,050 lb capacity
  • 0.172 in maximum deflection
  • 48 in width
  • 2.75 in overall height
  • Orange color
  • Made in the United States