Learn More
MSC Interlake Mecalux Pallet Rack Beam, 96 in OAW, 5 in OAH
Catalog No. 5000516952
Encompass
These slotted pallet rack beams are the horizontal elements of a pallet rack system where loads are placed. They are constructed from a single roll-formed piece of steel with a continuous seam weld on its base. Each load level in the system is formed by two beams, with one beam positioned at each post of the frame.
  • Roll-formed beams with welded end plates
  • End plates slot into the face of the tear drop upright frame
  • Beams are held in place with two safety, spring-loaded locking devices per beam
  • Piston lock frame-to-beam connection seats and locks automatically, protecting against accidental beam displacement
  • Rack Manufacturers Institute, Inc. (RMI) certified
Catalog No. 50-005-16952 Supplier MSC Supplier No. 09135062
These slotted pallet rack beams are the horizontal elements of a pallet rack system where loads are placed. They are constructed from a single roll-formed piece of steel with a continuous seam weld on its base. Each load level in the system is formed by two beams, with one beam positioned at each post of the frame.
  • Roll-formed beams with welded end plates
  • End plates slot into the face of the tear drop upright frame
  • Beams are held in place with two safety, spring-loaded locking devices per beam
  • Piston lock frame-to-beam connection seats and locks automatically, protecting against accidental beam displacement
  • Rack Manufacturers Institute, Inc. (RMI) certified