Learn More
Biomatik Corporation Interleukin 1 Alpha (Il1A) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29162
Encompass_Preferred
Interleukin 1 Alpha (Il1A) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Interleukin 1 Alpha (IL1a). The Interleukin 1 Alpha (Il1A) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Ser113~Ala271. Applications: WB, IHC. Target Synonyms: Interleukin 1 Alpha (IL1a). Accession Number: Q53QF9. Gene ID: 3552. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7564