Learn More
Creative Biomart Interleukin-10 (IL-10) Protein
Catalog No. 502084284
Encompass_Preferred
Recombinant human IL-10 is a non-glycosylated protein, containing 161 amino acids and having a molecular mass of 16.6 kDa.
Catalog No. 50-208-4284 Supplier Creative Biomart Supplier No. THP0233
Recombinant human IL-10 is a non-glycosylated protein, containing 161 amino acids and having a molecular mass of 16.6 kDa.