Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-10 receptor beta protein, rat, Fc fusion protein (HEK293) | >95.0% | 50 UG
SDP

Catalog No. 5000290382
Encompass_Preferred

Recombinant rat interleukin-10 receptor beta extracellular domain expressed in HEK293 cells and supplied as a C-terminal Fc fusion in lyophilized form. Suited for immunology research, receptor-ligand interaction studies, and assay development; provided as a 50 μg vial with high purity and low endotoxin.

  • Expression system: HEK293 mammalian cells.
  • Sequence length: 351 amino acids (M1-V351).
  • Molecular weight: approximately 68 kDa.
  • Tag: C-terminal Fc fusion (human Fc).
  • Format: lyophilized powder, formulated in PBS pH 7.4.
  • Quantity: 50 μg per vial.
  • Purity: >95% by reducing SDS-PAGE.
  • Endotoxin: <1 EU/μg (LAL).
  • Storage: store at -20°C; aliquot and freeze for long-term storage.

Catalog No. 50-002-90382 Supplier Medchemexpress LLC Supplier No. HYP7641450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat interleukin-10 receptor beta extracellular domain expressed in HEK293 cells and supplied as a C-terminal Fc fusion in lyophilized form. Suited for immunology research, receptor-ligand interaction studies, and assay development; provided as a 50 μg vial with high purity and low endotoxin.

  • Expression system: HEK293 mammalian cells.
  • Sequence length: 351 amino acids (M1-V351).
  • Molecular weight: approximately 68 kDa.
  • Tag: C-terminal Fc fusion (human Fc).
  • Format: lyophilized powder, formulated in PBS pH 7.4.
  • Quantity: 50 μg per vial.
  • Purity: >95% by reducing SDS-PAGE.
  • Endotoxin: <1 EU/μg (LAL).
  • Storage: store at -20°C; aliquot and freeze for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.