Learn More
Medchemexpress LLC Interleukin-10 receptor subunit beta | >95.0% | 50 UG
SDP
Catalog No. 5000293718
Encompass_Preferred
IL-10R beta protein is a recombinant human interleukin-10 receptor subunit produced in HEK293 cells with a C-terminal His-hFc tag for detection and purification. It is supplied for research use in studies of cytokine signaling and receptor interactions, and is characterized by high purity and low endotoxin for in vitro assays.
  • Recombinant human interleukin-10 receptor subunit beta produced in mammalian cells.
  • C-terminal His-hFc tag for easy detection and purification.
  • Expressed in HEK293 cells to support native folding and glycosylation.
  • High purity as determined by SDS-PAGE, suitable for biochemical assays.
  • Intended for research use only; not for human therapeutic use.
  • Available in small research-friendly quantities (50 μg).
Catalog No. 50-002-93718 Supplier Medchemexpress LLC Supplier No. HYP7483950UG
IL-10R beta protein is a recombinant human interleukin-10 receptor subunit produced in HEK293 cells with a C-terminal His-hFc tag for detection and purification. It is supplied for research use in studies of cytokine signaling and receptor interactions, and is characterized by high purity and low endotoxin for in vitro assays.
  • Recombinant human interleukin-10 receptor subunit beta produced in mammalian cells.
  • C-terminal His-hFc tag for easy detection and purification.
  • Expressed in HEK293 cells to support native folding and glycosylation.
  • High purity as determined by SDS-PAGE, suitable for biochemical assays.
  • Intended for research use only; not for human therapeutic use.
  • Available in small research-friendly quantities (50 μg).