Learn More
Medchemexpress LLC Interleukin-10 receptor subunit beta, recombinant human | 100 UG
SDP
Catalog No. 5000291589
Encompass_Preferred
Recombinant human interleukin-10 receptor subunit beta (IL-10Rβ) produced in HEK293 cells as a His-hFc fusion protein. Supplied as a purified, ready-to-use protein for receptor-binding studies, biochemical assays, and cell signaling experiments. Provided in small aliquots suitable for research applications.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • His-hFc fusion tag facilitates detection and capture.
  • Suitable for receptor binding assays, ELISA, and signaling studies.
  • Available in small research pack sizes (for example, 10 μg, 50 μg, 100 μg).
  • Store frozen to preserve activity and stability.
Catalog No. 50-002-91589 Supplier Medchemexpress LLC Supplier No. HYP74839100UG
Recombinant human interleukin-10 receptor subunit beta (IL-10Rβ) produced in HEK293 cells as a His-hFc fusion protein. Supplied as a purified, ready-to-use protein for receptor-binding studies, biochemical assays, and cell signaling experiments. Provided in small aliquots suitable for research applications.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • His-hFc fusion tag facilitates detection and capture.
  • Suitable for receptor binding assays, ELISA, and signaling studies.
  • Available in small research pack sizes (for example, 10 μg, 50 μg, 100 μg).
  • Store frozen to preserve activity and stability.