Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-12 beta protein (rabbit) | >95.0% | 100 UG
SDP

Catalog No. 5000293623
Encompass_Preferred

Recombinant rabbit interleukin-12 beta (p40) produced in HEK293 cells with a C-terminal 6xHis tag. Supplied as a lyophilized sterile powder for research applications; functions as the p40 subunit common to IL-12 and IL-23 and is suitable for biochemical assays and antibody generation.

  • Produced in HEK293 cells with C-terminal 6xHis tag.
  • Full-length mature protein (I23-N324), glycosylated.
  • Predicted molecular weight 36.1 kDa; observed ~40-50 kDa due to glycosylation.
  • Purity >95.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL assay).
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4; sterile powder.
  • Store at -20°C; add carrier protein for long-term storage.
  • Do not reconstitute below 100 μg/mL; freeze aliquots to minimize freeze-thaw.

Catalog No. 50-002-93623 Supplier Medchemexpress LLC Supplier No. HYP74836100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rabbit interleukin-12 beta (p40) produced in HEK293 cells with a C-terminal 6xHis tag. Supplied as a lyophilized sterile powder for research applications; functions as the p40 subunit common to IL-12 and IL-23 and is suitable for biochemical assays and antibody generation.

  • Produced in HEK293 cells with C-terminal 6xHis tag.
  • Full-length mature protein (I23-N324), glycosylated.
  • Predicted molecular weight 36.1 kDa; observed ~40-50 kDa due to glycosylation.
  • Purity >95.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg (LAL assay).
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4; sterile powder.
  • Store at -20°C; add carrier protein for long-term storage.
  • Do not reconstitute below 100 μg/mL; freeze aliquots to minimize freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.