Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-12 receptor subunit beta-2 | >95.0% | 10 UG
SDP

Catalog No. 5000293721
Encompass_Preferred

Recombinant IL-12R beta 2 protein (mouse) expressed in HEK293 cells with a C-terminal His tag, provided as a lyophilized sterile powder for biochemical and binding assays. The protein corresponds to amino acids N24-N637 and is characterized for purity and functional binding activity suitable for receptor-ligand studies.

  • Expressed in HEK293 cells with C-terminal His tag.
  • Amino acid range N24-N637.
  • Molecular weight approximately 100-130 kDa.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized sterile powder with stabilizers for storage.
  • Activity confirmed by ELISA (EC50 ≈ 19.97 ng/mL when immobilized at 0.5 μg/mL).
  • Store frozen; reconstituted material stable short term at 4°C.

Catalog No. 50-002-93721 Supplier Medchemexpress LLC Supplier No. HYP7483410UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant IL-12R beta 2 protein (mouse) expressed in HEK293 cells with a C-terminal His tag, provided as a lyophilized sterile powder for biochemical and binding assays. The protein corresponds to amino acids N24-N637 and is characterized for purity and functional binding activity suitable for receptor-ligand studies.

  • Expressed in HEK293 cells with C-terminal His tag.
  • Amino acid range N24-N637.
  • Molecular weight approximately 100-130 kDa.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized sterile powder with stabilizers for storage.
  • Activity confirmed by ELISA (EC50 ≈ 19.97 ng/mL when immobilized at 0.5 μg/mL).
  • Store frozen; reconstituted material stable short term at 4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.