Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-12 subunit beta | >95.0% | 2 UG
SDP

Catalog No. 5000293622
Encompass_Preferred

Recombinant rabbit interleukin-12 subunit beta (IL-12 p40) is a C-terminal His-tagged protein produced in HEK293 cells and supplied as a sterile, lyophilized powder. It is intended as a research reagent for immunology and cell signaling studies, including binding and functional assays involving IL-12/IL-23 pathways.

  • Recombinant rabbit IL-12 p40 produced in HEK293 cells.
  • C-terminal 6xHis tag for purification and detection.
  • Sterile lyophilized powder formulation for stability.
  • High purity, >95% by reducing SDS-PAGE.
  • Low endotoxin, <1 EU/μg, suitable for sensitive assays.
  • Glycosylated protein with apparent molecular weight ~40-50 kDa.
  • Functional activity with ED50 ≈5.816 ng/mL in receptor binding assay.
  • Store at -20 °C; reconstitute to ≥100 μg/mL and aliquot to avoid freeze-thaw.

Catalog No. 50-002-93622 Supplier Medchemexpress LLC Supplier No. HYP748362UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rabbit interleukin-12 subunit beta (IL-12 p40) is a C-terminal His-tagged protein produced in HEK293 cells and supplied as a sterile, lyophilized powder. It is intended as a research reagent for immunology and cell signaling studies, including binding and functional assays involving IL-12/IL-23 pathways.

  • Recombinant rabbit IL-12 p40 produced in HEK293 cells.
  • C-terminal 6xHis tag for purification and detection.
  • Sterile lyophilized powder formulation for stability.
  • High purity, >95% by reducing SDS-PAGE.
  • Low endotoxin, <1 EU/μg, suitable for sensitive assays.
  • Glycosylated protein with apparent molecular weight ~40-50 kDa.
  • Functional activity with ED50 ≈5.816 ng/mL in receptor binding assay.
  • Store at -20 °C; reconstitute to ≥100 μg/mL and aliquot to avoid freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.