Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-12 subunit beta | >95.0% | 50 UG
SDP

Catalog No. 5000292787
Encompass_Preferred

Recombinant interleukin-12 subunit beta (p40) derived from rabbit and produced in HEK293 cells. Supplied as a lyophilized powder (50 μg) with a C-terminal 6xHis tag and documented biological activity. Intended for research applications in immunology and cell signaling; store frozen and aliquot after reconstitution to preserve activity.

  • Produced in HEK293 cells with C-terminal 6xHis tag.
  • Supplied as lyophilized powder, easy to reconstitute.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Glycosylated; apparent molecular weight ~40-50 kDa.
  • Low endotoxin level (<1 EU/μg) suitable for many in vitro assays.
  • Recommended storage at -20 °C; aliquot and freeze at -80 °C for long-term.

Catalog No. 50-002-92787 Supplier Medchemexpress LLC Supplier No. HYP7483650UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant interleukin-12 subunit beta (p40) derived from rabbit and produced in HEK293 cells. Supplied as a lyophilized powder (50 μg) with a C-terminal 6xHis tag and documented biological activity. Intended for research applications in immunology and cell signaling; store frozen and aliquot after reconstitution to preserve activity.

  • Produced in HEK293 cells with C-terminal 6xHis tag.
  • Supplied as lyophilized powder, easy to reconstitute.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Glycosylated; apparent molecular weight ~40-50 kDa.
  • Low endotoxin level (<1 EU/μg) suitable for many in vitro assays.
  • Recommended storage at -20 °C; aliquot and freeze at -80 °C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.