Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-12 subunit beta (IL-12 p40) | 98.0% | 10 UG
SDP

Catalog No. 5000291537
Encompass_Preferred

Recombinant interleukin-12 subunit beta (IL-12 p40) derived from rabbit sequence and expressed in HEK293 cells with a C-terminal His tag. Supplied as a purified recombinant protein for research applications including immunology assays, signaling pathway studies, and antibody validation.

  • Recombinant protein produced in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity (98.0%) suitable for sensitive assays.
  • Glycosylated, apparent molecular weight ~40-50 kDa on SDS-PAGE.
  • Available in research-scale quantities (for example, 10 μg).
  • Store at -20 °C; shipped on blue ice to maintain stability.

Catalog No. 50-002-91537 Supplier Medchemexpress LLC Supplier No. HYP7483610UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant interleukin-12 subunit beta (IL-12 p40) derived from rabbit sequence and expressed in HEK293 cells with a C-terminal His tag. Supplied as a purified recombinant protein for research applications including immunology assays, signaling pathway studies, and antibody validation.

  • Recombinant protein produced in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity (98.0%) suitable for sensitive assays.
  • Glycosylated, apparent molecular weight ~40-50 kDa on SDS-PAGE.
  • Available in research-scale quantities (for example, 10 μg).
  • Store at -20 °C; shipped on blue ice to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.